Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760465.1 | XP_008758687.1 | 938 | |||
XM_008760466.1 | XP_008758688.1 | 938 | ||||
XM_008760467.1 | XP_008758689.1 | 938 | ||||
XM_008760468.1 | XP_008758690.1 | 529 |
Gene Symbol: LOC103690185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760460.1 | XP_008758682.1 | 8 | 938 | ||
XM_008760461.1 | XP_008758683.1 | 8 | 938 | |||
XM_008760462.1 | XP_008758684.1 | 8 | 938 | |||
XM_008760463.1 | XP_008758685.1 | 5 | 529 |