Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017190.4 | NP_058886.1 | 5 | 329 | ||
XM_006228824.2 | XP_006228886.1 | 4 | 359 | |||
XM_006228825.2 | XP_006228887.1 | 4 | 876 | |||
XM_006228827.2 | XP_006228889.1 | 5 | 411 | |||
XM_008759137.1 | XP_008757359.1 | 4 | 883 | |||
XM_008759138.1 | XP_008757360.1 | 4 | 352 | |||
XM_008759139.1 | XP_008757361.1 | 4 | 882 | |||
GenBank mRNA | FQ212027.1 | - | 5 | 345 | ||
M14871.1 | - | 4 | 299 | |||
M16800.1 | - | 5 | 329 | |||
M22357.1 | - | 4 | 277 | |||
X05301.1 | - | 4 | 292 |