Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133287.1 | NP_579821.1 | 2 | 145 | ||
GenBank mRNA | AB210108.1 | - | 2 | 145 | ||
BC166596.1 | - | 444 | 7934724 | |||
DQ336702.1 | - | 2 | 146 | |||
DQ336703.1 | - | 2 | 146 | |||
DQ336704.1 | - | 2 | 146 | |||
DQ336705.1 | - | 2 | 146 |