Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105727.2 | NP_001099197.2 | 2 | 491 | ||
GenBank mRNA | AI602737.1 | - | 2 | 372 | ||
D45247.1 | - | 2 | 489 | |||
FQ214283.1 | - | 1 | 25 |