Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772932.1 | XP_008771154.1 | 11, 12 | 1286 | ||
XM_008772933.1 | XP_008771155.1 | 10, 11 | 1148 | |||
GenBank mRNA | DQ054787.1 | - | 1 | 16 |