Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017200.1 | NP_058896.1 | 7 | 822 | ||
XM_006234440.2 | XP_006234502.1 | 8 | 1117 | |||
XM_006234441.2 | XP_006234503.1 | 8 | 2138 | |||
XM_006234444.2 | XP_006234506.1 | 8 | 2151 | |||
XM_008761980.1 | XP_008760202.1 | 8 | 1136 | |||
XM_008761981.1 | XP_008760203.1 | 8 | 900 | |||
XM_008761982.1 | XP_008760204.1 | 8 | 1130 | |||
XM_008761983.1 | XP_008760205.1 | 8 | 2153 | |||
XM_008761984.1 | XP_008760206.1 | 8 | 2166 | |||
GenBank mRNA | D10926.1 | - | 7 | 822 | ||
FQ219876.1 | - | 5 | 528 | |||
FQ220339.1 | - | 5 | 522 | |||
FQ220655.1 | - | 5 | 619 | |||
FQ229364.1 | - | 5 | 551 |