Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025114.1 | NP_001020285.1 | 4 | 353 | ||
XM_006256586.2 | XP_006256648.1 | 4 | 546 | |||
XM_006256587.2 | XP_006256649.1 | 4 | 547 | |||
GenBank mRNA | BC091322.1 | - | 4 | 353 | 7313558 |