Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106425.1 | NP_001099895.1 | 7 | 926 | ||
XM_008760864.1 | XP_008759086.1 | 7 | 935 | |||
GenBank mRNA | BC158721.1 | - | 7 | 917 | 9090461 |