Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019621.1 | NP_062567.1 | 12 | 1385 | ||
XM_006246680.2 | XP_006246742.1 | 14 | 1761 | |||
XM_006246681.2 | XP_006246743.1 | 14 | 1756 | |||
XM_006246682.2 | XP_006246744.1 | 14 | 1610 | |||
XM_006246683.2 | XP_006246745.1 | 14 | 1518 | |||
XM_006246684.2 | XP_006246746.1 | 14 | 1509 | |||
XM_006246685.2 | XP_006246747.1 | 12 | 1471 | |||
XM_006246686.2 | XP_006246748.1 | 12 | 1531 | |||
XM_006246687.2 | XP_006246749.1 | 12 | 1431 | |||
XM_006246688.2 | XP_006246750.1 | 11 | 3632 | |||
XM_006246689.2 | XP_006246751.1 | 11 | 1374 | |||
XM_006246691.2 | XP_006246753.1 | 11 | 1365 | |||
XM_008767809.1 | XP_008766031.1 | 14 | 1862 | |||
XM_008767810.1 | XP_008766032.1 | 11 | 1249 | |||
GenBank mRNA | M96853.1 | - | 12 | 1385 | ||
X66474.1 | - | 12 | 1530 |