Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761263.1 | XP_008759485.1 | 3 | 265 | ||
XM_227367.5 | XP_227367.5 | 2 | 308 |