Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106448.2 | NP_001099918.1 | 6, 7 | 913 | ||
GenBank mRNA | BC166534.1 | - | 6, 7 | 913 | 7190922 |