Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chi3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191712.1 | NP_001178641.1 | 5 | 378 | ||
XM_008761383.1 | XP_008759605.1 | 5 | 378 | |||
GenBank mRNA | JF781276.1 | - | 5 | 396 |