Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032416.1 | NP_115792.1 | 6 | 646 | ||
XM_006249385.2 | XP_006249447.1 | 6 | 710 | |||
GenBank mRNA | AF529165.2 | - | 6 | 523 | ||
AY566467.1 | - | 6 | 590 | |||
AY566468.1 | - | 6 | 590 | |||
AY566469.1 | - | 6 | 590 | |||
BC062081.1 | - | 6 | 622 | 5621229 | ||
X14977.1 | - | 6 | 646 |