Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013076.1 | NP_001013094.1 | 5 | 933 | ||
XM_006233488.1 | XP_006233550.1 | 4 | 915 | |||
XM_008761550.1 | XP_008759772.1 | 2 | 1300 | |||
GenBank mRNA | BC083638.1 | - | 5 | 933 | 7134832 |