Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199092.1 | NP_954523.1 | 14 | 1281 | ||
XM_006234150.2 | XP_006234212.1 | 16 | 1445 | |||
XM_006234151.2 | XP_006234213.1 | 15 | 1371 | |||
XM_006234152.2 | XP_006234214.1 | 17 | 1541 | |||
XM_006234154.2 | XP_006234216.1 | 13 | 1449 | |||
XM_008761704.1 | XP_008759926.1 | 14 | 1623 | |||
XM_008761705.1 | XP_008759927.1 | 16 | 1423 | |||
XM_008761706.1 | XP_008759928.1 | 15 | 1349 | |||
XM_008761707.1 | XP_008759929.1 | 15 | 1741 | |||
XM_008761708.1 | XP_008759930.1 | 14 | 1175 | |||
GenBank mRNA | BC060516.1 | - | 14 | 1281 | 5598062 | |
FQ213311.1 | - | 1231 | ||||
FQ230805.1 | - | 9 | 850 | |||
FQ231958.1 | - | 9 | 849 |