Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013916.1 | NP_001013938.1 | 3 | 498 | ||
GenBank mRNA | BC089829.1 | - | 3 | 498 | 7322853 | |
FQ209402.1 | - | 3 | 274 | |||
FQ213169.1 | - | 3 | 292 | |||
FQ215218.1 | - | 3 | 280 | |||
FQ216383.1 | - | 3 | 280 | |||
FQ219212.1 | - | 3 | 305 | |||
FQ219651.1 | - | 3 | 278 | |||
FQ219714.1 | - | 3 | 286 |