Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106496.1 | NP_001099966.1 | 5 | 526 | ||
XM_006234705.1 | XP_006234767.1 | 5 | 711 | |||
GenBank mRNA | FQ212852.1 | - | 5 | 678 | ||
FQ214203.1 | - | 5 | 678 |