Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011958.1 | NP_001011958.1 | 3 | 231 | ||
XM_008762314.1 | XP_008760536.1 | 3 | 621 | |||
XM_008762315.1 | XP_008760537.1 | 2 | 339 | |||
XM_008762316.1 | XP_008760538.1 | 3 | 299 | |||
XM_008762317.1 | XP_008760539.1 | 2 | 335 | |||
GenBank mRNA | BC087074.1 | - | 3 | 231 | 7134486 |