Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024870.1 | NP_001020041.1 | 2 | 128 | ||
GenBank mRNA | BC097352.1 | - | 2 | 128 | 7387542 |