Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106537.1 | NP_001100007.1 | 5 | 1100 | ||
XM_006235529.2 | XP_006235591.1 | 5 | 1152 |