Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134447.1 | NP_001127919.1 | 7, 8 | 881 | ||
GenBank mRNA | BC167106.1 | - | 7, 8 | 881 | 7133920 |