Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017274.1 | NP_058970.1 | 16 | 2028 | ||
GenBank mRNA | AF021348.1 | - | 16 | 2028 | ||
U36771.2 | - | 15 | 1801 |