Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017284.1 | NP_058980.1 | 1 | 127 | ||
GenBank mRNA | BC058487.1 | - | 1 | 147 | 6891616 | |
D21799.1 | - | 1 | 127 | |||
FQ217737.1 | - | 1 | 157 |