Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031597.4 | NP_113785.3 | 10 | 1777 | ||
XM_008765495.1 | XP_008763717.1 | 10 | 1105 | |||
GenBank mRNA | AF091247.1 | - | 11 | 1323 |