Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110139.2 | NP_001103609.1 | 16, 17 | 3018 | ||
NM_001110823.2 | NP_001104293.1 | 16, 17 | 3018 | |||
GenBank mRNA | AF043106.1 | - | 11, 12 | 1990 | ||
CK357706.1 | - | 3, 4 | 618 | |||
CK358684.1 | - | 1, 2 | 203 | |||
EV765256.1 | - | 2, 3 | 271 | |||
J04022.1 | - | 16, 17 | 3016 | |||
J04023.1 | - | 16, 17 | 2651 | |||
J04024.1 | - | 16, 17 | 2750 | |||
M25267.1 | - | 5, 6 | 1051 | |||
X15635.1 | - | 16, 17 | 2719 |