Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013083.2 | NP_001013101.2 | 11 | 1187 | ||
GenBank mRNA | CF249156.1 | - | 3 | 284 | ||
CF249511.1 | - | 5 | 551 | |||
FQ226028.1 | - | 8 | 870 | |||
FQ234275.1 | - | 7 | 736 |