Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019267.1 | NP_062140.1 | 4, 5 | 493 | ||
XM_006243450.2 | XP_006243512.1 | 5, 6 | 550 | |||
GenBank mRNA | BC158861.1 | - | 4, 5 | 486 | 7932093 | |
CK473898.1 | - | 4, 5 | 493 | |||
FQ213896.1 | - | 4, 5 | 475 | |||
FQ226240.1 | - | 4, 5 | 476 | |||
M94040.1 | - | 4, 5 | 404 |