Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017298.1 | NP_058994.1 | 33 | 4461 | ||
XM_006252586.2 | XP_006252648.1 | 34 | 4532 | |||
XM_006252590.2 | XP_006252652.1 | 33 | 4451 | |||
XM_006252594.2 | XP_006252656.1 | 32 | 4391 | |||
XM_006252595.2 | XP_006252657.1 | 31 | 4346 | |||
XM_006252597.2 | XP_006252659.1 | 31 | 4346 | |||
XM_008770990.1 | XP_008769212.1 | 25 | 5394 | |||
XM_008770991.1 | XP_008769213.1 | 32 | 4406 | |||
XM_008770992.1 | XP_008769214.1 | 32 | 4487 | |||
GenBank mRNA | AF370009.1 | - | 32 | 3963 | ||
AF422932.1 | - | 4 | 402 | |||
D38101.1 | - | 33 | 4461 | |||
D38102.1 | - | 33 | 4461 | |||
M57682.1 | - | 32 | 4498 | |||
M99221.1 | - | 6 | 727 | |||
U31772.1 | - | 5 | 457 |