Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017298.1 | NP_058994.1 | 49 | 6756 | ||
XM_006252586.2 | XP_006252648.1 | 50 | 6827 | |||
XM_006252590.2 | XP_006252652.1 | 49 | 6746 | |||
XM_006252594.2 | XP_006252656.1 | 48 | 6686 | |||
XM_006252595.2 | XP_006252657.1 | 47 | 6641 | |||
XM_006252597.2 | XP_006252659.1 | 46 | 6614 | |||
XM_008770990.1 | XP_008769212.1 | 41 | 7689 | |||
GenBank mRNA | AF370010.1 | - | 13 | 1878 | ||
D38101.1 | - | 49 | 6756 | |||
D38102.1 | - | 49 | 6698 |