Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034131.1 | NP_001029303.1 | 11, 12 | 1544 | ||
XM_006236954.2 | XP_006237016.1 | 10, 11 | 1270 | |||
XM_006236955.2 | XP_006237017.1 | 10, 11 | 1785 | |||
XM_006236956.1 | XP_006237018.1 | 10, 11 | 1207 | |||
XM_008763148.1 | XP_008761370.1 | 13, 14 | 2286 | |||
XM_008763149.1 | XP_008761371.1 | 13, 14 | 1688 | |||
XM_008763150.1 | XP_008761372.1 | 14, 15 | 1763 | |||
XM_008763151.1 | XP_008761373.1 | 13, 14 | 1880 | |||
XM_008763152.1 | XP_008761374.1 | 12, 13 | 1866 | |||
XM_008763153.1 | XP_008761375.1 | 12, 13 | 2412 | |||
XM_008763154.1 | XP_008761376.1 | 13, 14 | 2283 | |||
XM_008763155.1 | XP_008761377.1 | 13, 14 | 2284 | |||
XM_008763156.1 | XP_008761378.1 | 13, 14 | 2284 | |||
XM_008763157.1 | XP_008761379.1 | 13, 14 | 1682 | |||
XM_008763158.1 | XP_008761380.1 | 13, 14 | 1688 | |||
GenBank mRNA | BC100267.1 | - | 11, 12 | 1544 | 7383433 |