Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034131.1 | NP_001029303.1 | 1 | 102 | ||
XM_008763148.1 | XP_008761370.1 | 3 | 844 | |||
XM_008763149.1 | XP_008761371.1 | 3 | 246 | |||
XM_008763150.1 | XP_008761372.1 | 4 | 321 | |||
XM_008763151.1 | XP_008761373.1 | 3 | 438 | |||
XM_008763152.1 | XP_008761374.1 | 2 | 424 | |||
XM_008763153.1 | XP_008761375.1 | 2 | 970 | |||
XM_008763154.1 | XP_008761376.1 | 3 | 844 | |||
XM_008763155.1 | XP_008761377.1 | 3 | 845 | |||
XM_008763156.1 | XP_008761378.1 | 3 | 845 | |||
XM_008763157.1 | XP_008761379.1 | 3 | 246 | |||
XM_008763158.1 | XP_008761380.1 | 3 | 246 | |||
GenBank mRNA | BC100267.1 | - | 1 | 102 | 7383433 |