Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017309.2 | NP_059005.1 | 2 | 232 | ||
XM_006251517.2 | XP_006251579.2 | 2 | 275 | |||
GenBank mRNA | BC088855.1 | - | 2 | 232 | 7317429 | |
D14425.1 | - | 2 | 200 | |||
D14568.1 | - | 2 | 88 | |||
FQ212066.1 | - | 2 | 399 | |||
L03554.1 | - | 1 | 34 |