Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019274.2 | NP_062147.1 | 1 | 64 | ||
GenBank mRNA | AF007583.1 | - | 1 | 64 |