Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC502894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025064.1 | NP_001020235.1 | 5 | 725 | ||
XM_006237501.2 | XP_006237563.1 | 6 | 804 | |||
GenBank mRNA | BC093610.1 | - | 5 | 725 | 7319640 | |
FQ217296.1 | - | 6 | 766 |