Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106635.1 | NP_001100105.1 | 13 | 2540 | ||
XM_006239011.2 | XP_006239073.1 | 14 | 3737 | |||
XM_006239012.2 | XP_006239074.1 | 14 | 3734 |