Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039914.2 | NP_001035003.1 | 1 | 236 | ||
XM_008763614.1 | XP_008761836.1 | 1 | 483 | |||
GenBank mRNA | AB234867.1 | - | 1 | 256 |