Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011974.1 | NP_001011974.1 | 2 | 1977 | ||
XM_006238184.2 | XP_006238246.1 | 2 | 2542 | |||
XM_006238185.2 | XP_006238247.1 | 2 | 1959 | |||
XM_006238186.2 | XP_006238248.1 | 2 | 2058 | |||
XM_008763733.1 | XP_008761955.1 | 3 | 2024 | |||
GenBank mRNA | BC085790.1 | - | 2 | 1977 | 7108964 |