Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100965.2 | NP_001094435.1 | 2 | 794 | ||
XM_008763860.1 | XP_008762082.1 | 4 | 1053 | |||
XM_008763861.1 | XP_008762083.1 | 3 | 982 | |||
XM_008763862.1 | XP_008762084.1 | 4 | 954 | |||
XM_008763863.1 | XP_008762085.1 | 4 | 1336 | |||
XM_008763864.1 | XP_008762086.1 | 3 | 883 | |||
XM_008763865.1 | XP_008762087.1 | 3 | 811 | |||
XM_008763866.1 | XP_008762088.1 | 4 | 1125 | |||
GenBank mRNA | BC133066.1 | - | 2 | 797 | 7134332 |