Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069267.4 | XP_001069267.2 | 7 | 1058 | ||
XM_008763922.1 | XP_008762144.1 | 7 | 1047 | |||
XM_008763923.1 | XP_008762145.1 | 7 | 1048 | |||
XM_008763924.1 | XP_008762146.1 | 7 | 1046 | |||
XM_008763925.1 | XP_008762147.1 | 7 | 1034 | |||
XM_008763926.1 | XP_008762148.1 | 7 | 1038 | |||
XM_008763927.1 | XP_008762149.1 | 7 | 1042 | |||
XM_008763928.1 | XP_008762150.1 | 6 | 981 | |||
XM_008763929.1 | XP_008762151.1 | 6 | 954 | |||
XM_008763930.1 | XP_008762152.1 | 7 | 1158 |