Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106679.2 | NP_001100149.2 | 5 | 1066 | ||
XM_006238640.2 | XP_006238702.1 | 6 | 1238 | |||
XM_006238642.2 | XP_006238704.1 | 6 | 1249 | |||
XM_006238643.2 | XP_006238705.1 | 6 | 1250 | |||
XM_006238644.2 | XP_006238706.1 | 6 | 1250 | |||
XM_008763992.1 | XP_008762214.1 | 6 | 1249 | |||
GenBank mRNA | FQ225549.1 | - | 4 | 368 |