Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013935.1 | NP_001013957.1 | 6 | 586 | ||
XM_006239124.2 | XP_006239186.1 | 6 | 882 | |||
XM_006239125.2 | XP_006239187.1 | 6 | 660 | |||
GenBank mRNA | BC083692.1 | - | 6 | 586 | 7189448 |