Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024758.2 | NP_001019929.1 | 8, 9 | 879 | ||
NM_001170533.1 | NP_001164004.1 | 9, 10 | 1301 | |||
XM_006239559.2 | XP_006239621.1 | 9, 10 | 1461 | |||
XM_006239562.2 | XP_006239624.1 | 7, 8 | 1109 | |||
XM_008764356.1 | XP_008762578.1 | 10, 11 | 2748 | |||
XM_008764357.1 | XP_008762579.1 | 9, 10 | 1257 | |||
GenBank mRNA | BC083833.1 | - | 6, 7 | 718 | ||
BC097417.1 | - | 8, 9 | 879 | 7454407 |