Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106701.2 | NP_001100171.1 | 4 | 469 | ||
GenBank mRNA | BC158867.1 | - | 4 | 469 | 5623763 | |
FQ219403.1 | - | 3 | 877 |