Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013095.1 | NP_001013113.1 | 7, 8 | 593 | ||
GenBank mRNA | BC086383.1 | - | 7, 8 | 593 | 7301572 | |
FQ213005.1 | - | 7, 8 | 634 | |||
FQ225883.1 | - | 7, 8 | 626 | |||
FQ226276.1 | - | 7, 8 | 625 | |||
FQ229703.1 | - | 7, 8 | 624 | |||
FQ230888.1 | - | 7, 8 | 619 | |||
FQ232661.1 | - | 7, 8 | 623 | |||
FQ234018.1 | - | 7, 8 | 627 | |||
FQ234727.1 | - | 5, 6 | 475 | |||
FQ234921.1 | - | 7, 8 | 624 | |||
FQ234929.1 | - | 7, 8 | 626 | |||
FQ235264.1 | - | 4, 5 | 407 |