Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257347.1 | NP_001244276.1 | 11 | 1935 | ||
XM_006242585.2 | XP_006242647.1 | 13 | 2246 | |||
XM_006242586.2 | XP_006242648.1 | 13 | 2205 |