Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011981.1 | NP_001011981.1 | 15 | 1502 | ||
GenBank mRNA | BC078931.1 | - | 15 | 1502 | 7107232 |