Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106715.1 | NP_001100185.1 | 17 | 2616 | ||
XM_006239879.2 | XP_006239941.1 | 17 | 2712 | |||
XM_006239880.2 | XP_006239942.1 | 17 | 2706 | |||
XM_006239883.2 | XP_006239945.1 | 16 | 2469 | |||
XM_006239884.2 | XP_006239946.2 | 16 | 3388 | |||
XM_008764579.1 | XP_008762801.1 | 16 | 3381 | |||
XM_008764580.1 | XP_008762802.1 | 17 | 2733 | |||
XM_008764581.1 | XP_008762803.1 | 15 | 3150 | |||
XM_008764582.1 | XP_008762804.1 | 15 | 3143 | |||
GenBank mRNA | AY724506.1 | - | 15 | 2240 | ||
FQ226250.1 | - | 2 | 196 | |||
FQ226792.1 | - | 1 | 17 | |||
FQ232361.1 | - | 5 | 774 |