Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106726.1 | NP_001100196.1 | 11 | 1221 | ||
XM_006240078.2 | XP_006240140.1 | 10 | 1147 | |||
XM_006240079.2 | XP_006240141.1 | 12 | 1312 | |||
XM_006240080.2 | XP_006240142.1 | 11 | 2674 | |||
GenBank mRNA | FQ221669.1 | - | 12 | 2141 |