Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100542.1 | NP_001094012.1 | 10 | 1067 | ||
GenBank mRNA | BC091286.1 | - | 8 | 850 | ||
BC166783.1 | - | 10 | 1049 | 7935021 |