Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1305089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106730.2 | NP_001100200.1 | 1, 2 | 1495 | ||
GenBank mRNA | BC168878.1 | - | 1, 2 | 1495 | 7382020 |